CH4424851258 ChemBridge Corporation

1-(2-methoxybenzyl)piperazine bis(trifluoroacetate)

  • MDL number MFCD11841334
  • Cas Number : 1192656-95-3
  • Molecular Weight : 434.3308

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data 2CF3COOH
Storage temp. RT
Physical Form solid
Shipping Temperature RT
Country of Origin Russia
Product Link https://www.hit2lead.com/building-blocks/4103036

Safety Information

MSDS https://www.hit2lead.com/sds/4103036_sds.pdf
Availability details for
Quantity
  • Availability
  • Add to Cart