CH4424851263 ChemBridge Corporation

1-(3-methylbenzyl)piperazine bis(trifluoroacetate)

  • MDL number MFCD11841338
  • Cas Number : 1185491-33-1
  • Molecular Weight : 418.3314

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data 2CF3COOH
Storage temp. Room Temperature
Shipping Temperature Room Temperature
Country of Origin Russia
Product Link https://www.hit2lead.com/building-blocks/4103044

Safety Information

MSDS https://www.hit2lead.com/sds/4103044_sds.pdf
Availability details for
Quantity
  • Availability
  • Add to Cart