CH4424851264 ChemBridge Corporation

[2-(5-chloro-1H-benzimidazol-2-yl)ethyl]amine dihydrochloride

  • MDL number MFCD03419439
  • Cas Number : 91003-86-0
  • Molecular Weight : 268.5707

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data 2HCl
Storage temp. Room Temperature
Shipping Temperature Room Temperature
Country of Origin Russia
Product Link https://www.hit2lead.com/building-blocks/4110558

Safety Information

MSDS https://www.hit2lead.com/sds/4110558_sds.pdf
Availability details for
Quantity
  • Availability
  • Add to Cart