CH4424851286 ChemBridge Corporation

5-(aminomethyl)-2-furoic acid hydrochloride

  • MDL number MFCD20731136
  • Cas Number : 51521-95-0
  • Molecular Weight : 177.5856

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data HCl
Storage temp. RT
Physical Form solid
Shipping Temperature RT
Country of Origin Russia
Product Link https://www.hit2lead.com/building-blocks/4141271

Safety Information

MSDS https://www.hit2lead.com/sds/4141271_sds.pdf
Availability details for
Quantity
  • Availability
  • Add to Cart