CH4424851293 ChemBridge Corporation

2-methyl-D-proline hydrochloride

  • MDL number MFCD20487714
  • Cas Number : 123053-48-5
  • Molecular Weight : 165.618

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data HCl
Storage temp. Room Temperature
Shipping Temperature Room Temperature
Country of Origin Russia
Product Link https://www.hit2lead.com/building-blocks/4142037

Safety Information

MSDS https://www.hit2lead.com/sds/4142037_sds.pdf
Availability details for
Quantity
  • Availability
  • Add to Cart