CH4424851332 ChemBridge Corporation

(1-oxo-4-phenyl-2(1H)-phthalazinyl)acetic acid hydrate

  • MDL number MFCD13194045
  • Cas Number : 1609403-19-1
  • Molecular Weight : 298.2934

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data H2O
Storage temp. Room Temperature
Shipping Temperature Room Temperature
Country of Origin Russia
Product Link https://www.hit2lead.com/building-blocks/4400204

Safety Information

MSDS https://www.hit2lead.com/sds/4400204_sds.pdf
Availability details for
Quantity
  • Availability
  • Add to Cart