CH4424851373 ChemBridge Corporation

1-piperidinylacetic acid hydrate

  • MDL number MFCD07781088
  • Cas Number : 478920-83-1
  • Molecular Weight : 161.1989

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data H2O
Storage temp. RT
Physical Form solid
Shipping Temperature RT
Country of Origin Russia
Product Link https://www.hit2lead.com/building-blocks/4401702

Safety Information

MSDS https://www.hit2lead.com/sds/4401702_sds.pdf
Availability details for
Quantity
  • Availability
  • Add to Cart