CH4424851376 ChemBridge Corporation

(2-methyl-1H-benzimidazol-1-yl)acetic acid hydrochloride hydrate

  • MDL number MFCD26959583
  • Molecular Weight : 244.6748

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data HCl H2O
Storage temp. RT
Physical Form solid
Shipping Temperature RT
Country of Origin Russia
Product Link https://www.hit2lead.com/building-blocks/4401766

Safety Information

MSDS https://www.hit2lead.com/sds/4401766_sds.pdf
Availability details for
Quantity
  • Availability
  • Add to Cart