CH4424851388 ChemBridge Corporation

8-methylimidazo[1,2-a]pyridine-2-carboxylic acid hydrochloride hydrate

  • MDL number MFCD13194050
  • Molecular Weight : 230.6482

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data HCl H2O
Storage temp. Room Temperature
Shipping Temperature Room Temperature
Country of Origin Russia
Product Link https://www.hit2lead.com/building-blocks/4402937

Safety Information

MSDS https://www.hit2lead.com/sds/4402937_sds.pdf
Availability details for
Quantity
  • Availability
  • Add to Cart