CH4424851415 ChemBridge Corporation

N,3-dimethyltetrahydro-3-thiophenamine 1,1-dioxide hydrochloride

  • MDL number MFCD05885471
  • Cas Number : 5553-29-7
  • Molecular Weight : 199.6989

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data HCl
Storage temp. Room Temperature
Shipping Temperature Room Temperature
Country of Origin Russia
Product Link https://www.hit2lead.com/building-blocks/5155765

Safety Information

MSDS https://www.hit2lead.com/building-blocks/5155765
Availability details for
Quantity
  • Availability
  • Add to Cart