CH4424851422 ChemBridge Corporation

1-amino-3-(4-tert-butylphenoxy)-2-propanol hydrochloride

  • MDL number MFCD26959630
  • Cas Number : 1609396-18-0
  • Molecular Weight : 259.7723

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data HCl
Storage temp. Room Temperature
Shipping Temperature Room Temperature
Country of Origin Russia
Product Link https://www.hit2lead.com/building-blocks/5182179

Safety Information

MSDS https://www.hit2lead.com/sds/5182179_sds.pdf
Availability details for
Quantity
  • Availability
  • Add to Cart