CH4424851425 ChemBridge Corporation

1-(2-chloro-4-nitrophenyl)-3-methylpiperazine hydrochloride

  • MDL number MFCD00545969
  • Cas Number : 299425-75-5
  • Molecular Weight : 292.1617

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data HCl
Storage temp. Room Temperature
Shipping Temperature Room Temperature
Country of Origin Russia
Product Link https://www.hit2lead.com/building-blocks/5200236

Safety Information

MSDS https://www.hit2lead.com/sds/5200236_sds.pdf
Availability details for
Quantity
  • Availability
  • Add to Cart