CH4424851459 ChemBridge Corporation

N-(4-chlorobenzyl)-2-methyl-1-propanamine hydrochloride

  • MDL number MFCD13186367
  • Cas Number : 222846-44-8
  • Molecular Weight : 234.1654

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data HCl
Storage temp. RT
Physical Form solid
Shipping Temperature RT
Country of Origin Russia
Product Link https://www.hit2lead.com/building-blocks/5402655

Safety Information

MSDS https://www.hit2lead.com/sds/5402655_sds.pdf
Availability details for
Quantity
  • Availability
  • Add to Cart