CH4424851460 ChemBridge Corporation

N-(3-pyridinylmethyl)cyclohexanamine dihydrochloride

  • MDL number MFCD06071930
  • Cas Number : 1158310-28-1
  • Molecular Weight : 263.2066

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data 2HCl
Storage temp. RT
Physical Form solid
Shipping Temperature RT
Country of Origin Russia
Product Link https://www.hit2lead.com/building-blocks/5402674

Safety Information

MSDS https://www.hit2lead.com/sds/5402674_sds.pdf
Availability details for
Quantity
  • Availability
  • Add to Cart