CH4424851461 ChemBridge Corporation

2-methyl-N-(3-pyridinylmethyl)-1-propanamine dihydrochloride

  • MDL number MFCD13193984
  • Cas Number : 1240578-47-5
  • Molecular Weight : 237.1693

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data 2HCl
Storage temp. RT
Physical Form solid
Shipping Temperature RT
Country of Origin Russia
Product Link https://www.hit2lead.com/building-blocks/5402676

Safety Information

MSDS https://www.hit2lead.com/sds/5402676_sds.pdf
Availability details for
Quantity
  • Availability
  • Add to Cart