CH4424851463 ChemBridge Corporation

N-(4-fluorobenzyl)-2-propanamine hydrochloride

  • MDL number MFCD08706976
  • Cas Number : 1158308-41-8
  • Molecular Weight : 203.6842

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data HCl
Storage temp. RT
Physical Form solid
Shipping Temperature RT
Country of Origin Russia
Product Link https://www.hit2lead.com/building-blocks/5402692

Safety Information

MSDS https://www.hit2lead.com/sds/5402692_sds.pdf
Availability details for
Quantity
  • Availability
  • Add to Cart