CH4424851464 ChemBridge Corporation

N-(4-fluorobenzyl)cyclohexanamine hydrochloride

  • MDL number MFCD13186275
  • Cas Number : 1609396-07-7
  • Molecular Weight : 243.7481

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data HCl
Storage temp. RT
Physical Form solid
Shipping Temperature RT
Country of Origin Russia
Product Link https://www.hit2lead.com/building-blocks/5402694

Safety Information

MSDS https://www.hit2lead.com/sds/5402694_sds.pdf
Availability details for
Quantity
  • Availability
  • Add to Cart