CH4424851479 ChemBridge Corporation

3-chloro-N-3-pyridinylpropanamide hydrochloride

  • MDL number MFCD13193995
  • Cas Number : 1609407-51-3
  • Molecular Weight : 221.0838

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data HCl
Storage temp. RT
Physical Form solid
Shipping Temperature RT
Country of Origin Russia
Product Link https://www.hit2lead.com/building-blocks/5522444

Safety Information

MSDS https://www.hit2lead.com/sds/5522444_sds.pdf
Availability details for
Quantity
  • Availability
  • Add to Cart