CH4424851497 ChemBridge Corporation

N-(3,4-dimethoxybenzyl)-2-propanamine hydrochloride

  • MDL number MFCD05982316
  • Cas Number : 1049788-24-0
  • Molecular Weight : 245.7457

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data HCl
Storage temp. RT
Physical Form solid
Shipping Temperature RT
Country of Origin Russia
Product Link https://www.hit2lead.com/building-blocks/5525060

Safety Information

MSDS https://www.hit2lead.com/sds/5525060_sds.pdf
Availability details for
Quantity
  • Availability
  • Add to Cart