CH4424851507 ChemBridge Corporation

(2-methoxybenzyl)[2-(trifluoromethyl)benzyl]amine hydrobromide

  • MDL number MFCD13186388
  • Cas Number : 1609407-20-6
  • Molecular Weight : 376.2115

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data HBr
Storage temp. Room Temperature
Shipping Temperature Room Temperature
Country of Origin Russia
Product Link https://www.hit2lead.com/building-blocks/5527822

Safety Information

MSDS https://www.hit2lead.com/sds/5527822_sds.pdf
Availability details for
Quantity
  • Availability
  • Add to Cart