CH4424851509 ChemBridge Corporation

2-{2-[(2-hydroxyethyl)amino]-1H-benzimidazol-1-yl}-1-phenylethanone hydrobromide

  • MDL number MFCD00965590
  • Cas Number : 174312-21-1
  • Molecular Weight : 376.2477

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data HBr
Storage temp. RT
Physical Form solid
Shipping Temperature RT
Country of Origin Russia
Product Link https://www.hit2lead.com/building-blocks/5528440

Safety Information

MSDS https://www.hit2lead.com/sds/5528440_sds.pdf
Availability details for
Quantity
  • Availability
  • Add to Cart