CH4424851513 ChemBridge Corporation

(1,3-benzodioxol-5-ylmethyl)(4-pyridinylmethyl)amine dihydrobromide

  • MDL number MFCD26959641
  • Cas Number : 1609407-16-0
  • Molecular Weight : 404.097

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data 2HBr
Storage temp. RT
Physical Form solid
Shipping Temperature RT
Country of Origin Russia
Product Link https://www.hit2lead.com/building-blocks/5529896

Safety Information

MSDS https://www.hit2lead.com/sds/5529896_sds.pdf
Availability details for
Quantity
  • Availability
  • Add to Cart