CH4424851519 ChemBridge Corporation

(2-methoxy-1-methylethyl)[(4-methoxy-1-naphthyl)methyl]amine hydrobromide

  • MDL number MFCD13186395
  • Cas Number : 1609403-90-8
  • Molecular Weight : 340.2554

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data HBr
Storage temp. Room Temperature
Shipping Temperature Room Temperature
Country of Origin Russia
Product Link https://www.hit2lead.com/building-blocks/5530762

Safety Information

MSDS https://www.hit2lead.com/sds/5530762_sds.pdf
Availability details for
Quantity
  • Availability
  • Add to Cart