CH4424851522 ChemBridge Corporation

N-(2,4-dimethoxybenzyl)-3-pentanamine hydrobromide

  • MDL number MFCD13186396
  • Cas Number : 1609407-03-5
  • Molecular Weight : 318.24

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 0.95
Salt Data HBr
Storage temp. RT
Physical Form solid
Shipping Temperature RT
Country of Origin Russia
Product Link https://www.hit2lead.com/building-blocks/5531381

Safety Information

MSDS https://www.hit2lead.com/sds/5531381_sds.pdf
Availability details for
Quantity
  • Availability
  • Add to Cart