CH4424851523 ChemBridge Corporation

N-(3-bromo-4-methoxybenzyl)-2-(4-chlorophenyl)ethanamine hydrobromide

  • MDL number MFCD13186397
  • Cas Number : 1609403-93-1
  • Molecular Weight : 435.5812

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data HBr
Storage temp. Room Temperature
Shipping Temperature Room Temperature
Country of Origin Russia
Product Link https://www.hit2lead.com/building-blocks/5531840

Safety Information

MSDS https://www.hit2lead.com/sds/5531840_sds.pdf
Availability details for
Quantity
  • Availability
  • Add to Cart