CH4424851530 ChemBridge Corporation

N-(2-pyridinylmethyl)cyclohexanamine dihydrobromide

  • MDL number MFCD13193997
  • Cas Number : 1609403-47-5
  • Molecular Weight : 352.1086

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data 2HBr
Storage temp. RT
Physical Form solid
Shipping Temperature RT
Country of Origin Russia
Product Link https://www.hit2lead.com/building-blocks/5533964

Safety Information

MSDS https://www.hit2lead.com/sds/5533964_sds.pdf
Availability details for
Quantity
  • Availability
  • Add to Cart