CH4424851535 ChemBridge Corporation

N-(4-methoxybenzyl)-2-phenylethanamine hydrochloride

  • MDL number MFCD00034841
  • Molecular Weight : 277.7891

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data HCl
Storage temp. RT
Physical Form solid
Shipping Temperature RT
Country of Origin Russia
Product Link https://www.hit2lead.com/building-blocks/5534416

Safety Information

MSDS https://www.hit2lead.com/sds/5534416_sds.pdf
Availability details for
Quantity
  • Availability
  • Add to Cart