CH4424851541 ChemBridge Corporation

N-(2,4-dimethoxybenzyl)-2-(4-fluorophenyl)ethanamine hydrochloride

  • MDL number MFCD13186279
  • Cas Number : 1609407-33-1
  • Molecular Weight : 325.8055

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data HCl
Storage temp. RT
Physical Form solid
Shipping Temperature RT
Country of Origin Russia
Product Link https://www.hit2lead.com/building-blocks/5535937

Safety Information

MSDS https://www.hit2lead.com/sds/5535937_sds.pdf
Availability details for
Quantity
  • Availability
  • Add to Cart