CH4424851542 ChemBridge Corporation

N-(2,3-dimethoxybenzyl)-2-propanamine hydrochloride

  • MDL number MFCD08707173
  • Cas Number : 1158345-38-0
  • Molecular Weight : 245.7457

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data HCl
Storage temp. RT
Physical Form solid
Shipping Temperature RT
Country of Origin Russia
Product Link https://www.hit2lead.com/building-blocks/5536019

Safety Information

MSDS https://www.hit2lead.com/sds/5536019_sds.pdf
Availability details for
Quantity
  • Availability
  • Add to Cart