CH4424851544 ChemBridge Corporation

N-(2,4,5-trimethoxybenzyl)cyclohexanamine hydrobromide

  • MDL number MFCD13186281
  • Cas Number : 1609407-25-1
  • Molecular Weight : 360.2865

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data HBr
Storage temp. Room Temperature
Shipping Temperature Room Temperature
Country of Origin Russia
Product Link https://www.hit2lead.com/building-blocks/5536631

Safety Information

MSDS https://www.hit2lead.com/sds/5536631_sds.pdf
Availability details for
Quantity
  • Availability
  • Add to Cart