CH4424851546 ChemBridge Corporation

(4-methoxybenzyl)(2-nitrobenzyl)amine hydrobromide

  • MDL number MFCD13186250
  • Cas Number : 1609403-54-4
  • Molecular Weight : 353.2111

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data HBr
Storage temp. RT
Physical Form solid
Shipping Temperature RT
Country of Origin Russia
Product Link https://www.hit2lead.com/building-blocks/5536759

Safety Information

MSDS https://www.hit2lead.com/sds/5536759_sds.pdf
Availability details for
Quantity
  • Availability
  • Add to Cart