CH4424851550 ChemBridge Corporation

N-(2-nitrobenzyl)-2-phenylethanamine hydrobromide

  • MDL number MFCD13186254
  • Cas Number : 1609403-62-4
  • Molecular Weight : 337.2117

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data HBr
Storage temp. Room Temperature
Shipping Temperature Room Temperature
Country of Origin Russia
Product Link https://www.hit2lead.com/building-blocks/5536892

Safety Information

MSDS https://www.hit2lead.com/sds/5536892_sds.pdf
Availability details for
Quantity
  • Availability
  • Add to Cart