CH4424851555 ChemBridge Corporation

N-(3-nitrobenzyl)-3-pentanamine hydrobromide

  • MDL number MFCD13186259
  • Cas Number : 1609396-58-8
  • Molecular Weight : 303.1955

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data HBr
Storage temp. Room Temperature
Shipping Temperature Room Temperature
Country of Origin Russia
Product Link https://www.hit2lead.com/building-blocks/5537359

Safety Information

MSDS https://www.hit2lead.com/sds/5537359_sds.pdf
Availability details for
Quantity
  • Availability
  • Add to Cart