CH4424851562 ChemBridge Corporation

N-(4-methoxybenzyl)-2-(4-methoxyphenyl)ethanamine hydrobromide

  • MDL number MFCD13186403
  • Cas Number : 1609407-17-1
  • Molecular Weight : 352.2661

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data HBr
Storage temp. Room Temperature
Shipping Temperature Room Temperature
Country of Origin Russia
Product Link https://www.hit2lead.com/building-blocks/5539120

Safety Information

MSDS https://www.hit2lead.com/sds/5539120_sds.pdf
Availability details for
Quantity
  • Availability
  • Add to Cart