CH4424851568 ChemBridge Corporation

N-(3,4-dimethoxybenzyl)cyclopentanamine hydrochloride

  • MDL number MFCD06800487
  • Cas Number : 210291-83-1
  • Molecular Weight : 271.783

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data HCl
Storage temp. RT
Physical Form solid
Shipping Temperature RT
Country of Origin Russia
Product Link https://www.hit2lead.com/building-blocks/5540698

Safety Information

MSDS https://www.hit2lead.com/sds/5540698_sds.pdf
Availability details for
Quantity
  • Availability
  • Add to Cart