CH4424851570 ChemBridge Corporation

N-(2,4,5-trimethoxybenzyl)-3-pentanamine hydrobromide

  • MDL number MFCD13186297
  • Cas Number : 1609402-77-8
  • Molecular Weight : 348.2758

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data HBr
Storage temp. Room Temperature
Shipping Temperature Room Temperature
Country of Origin Russia
Product Link https://www.hit2lead.com/building-blocks/5540900

Safety Information

MSDS https://www.hit2lead.com/sds/5540900_sds.pdf
Availability details for
Quantity
  • Availability
  • Add to Cart