CH4424851577 ChemBridge Corporation

N-(2,4-dimethoxybenzyl)-1,2,3,4-tetrahydro-1-naphthalenamine hydrobromide

  • MDL number MFCD13186407
  • Cas Number : 1609407-35-3
  • Molecular Weight : 378.3034

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data HBr
Storage temp. Room Temperature
Shipping Temperature Room Temperature
Country of Origin Russia
Product Link https://www.hit2lead.com/building-blocks/5542161

Safety Information

MSDS https://www.hit2lead.com/sds/5542161_sds.pdf
Availability details for
Quantity
  • Availability
  • Add to Cart