CH4424851588 ChemBridge Corporation

N-(2,4-dimethoxybenzyl)-2-phenylethanamine hydrobromide

  • MDL number MFCD13186414
  • Cas Number : 1609396-31-7
  • Molecular Weight : 352.2661

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data HBr
Storage temp. RT
Physical Form solid
Shipping Temperature RT
Country of Origin Russia
Product Link https://www.hit2lead.com/building-blocks/5543528

Safety Information

MSDS https://www.hit2lead.com/sds/5543528_sds.pdf
Availability details for
Quantity
  • Availability
  • Add to Cart