CH4424851590 ChemBridge Corporation

N-(5-bromo-2-methoxybenzyl)-2-(1H-indol-3-yl)ethanamine hydrobromide

  • MDL number MFCD13186415
  • Cas Number : 1609407-64-8
  • Molecular Weight : 440.1722

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data HBr
Storage temp. RT
Physical Form solid
Shipping Temperature RT
Country of Origin Russia
Product Link https://www.hit2lead.com/building-blocks/5544480

Safety Information

MSDS https://www.hit2lead.com/sds/5544480_sds.pdf
Availability details for
Quantity
  • Availability
  • Add to Cart