CH4424851591 ChemBridge Corporation

N-(3,4-dimethoxybenzyl)-1-methoxy-2-propanamine hydrobromide

  • MDL number MFCD13186416
  • Cas Number : 1609396-34-0
  • Molecular Weight : 320.2227

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data HBr
Storage temp. Room Temperature
Shipping Temperature Room Temperature
Country of Origin Russia
Product Link https://www.hit2lead.com/building-blocks/5544614

Safety Information

MSDS https://www.hit2lead.com/sds/5544614_sds.pdf
Availability details for
Quantity
  • Availability
  • Add to Cart