CH4424851593 ChemBridge Corporation

N-(3-methoxybenzyl)ethanamine hydrochloride

  • MDL number MFCD07108408
  • Cas Number : 1048948-04-4
  • Molecular Weight : 201.6931

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data HCl
Storage temp. RT
Physical Form solid
Shipping Temperature RT
Country of Origin Russia
Product Link https://www.hit2lead.com/building-blocks/5545112

Safety Information

MSDS https://www.hit2lead.com/sds/5545112_sds.pdf
Availability details for
Quantity
  • Availability
  • Add to Cart