CH4424851594 ChemBridge Corporation

N-(3-ethoxy-4-methoxybenzyl)-3-pentanamine hydrobromide

  • MDL number MFCD13186418
  • Cas Number : 1609404-26-3
  • Molecular Weight : 332.2764

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data HBr
Storage temp. RT
Physical Form solid
Shipping Temperature RT
Country of Origin Russia
Product Link https://www.hit2lead.com/building-blocks/5545245

Safety Information

MSDS https://www.hit2lead.com/sds/5545245_sds.pdf
Availability details for
Quantity
  • Availability
  • Add to Cart