CH4424851595 ChemBridge Corporation

N-(2,4,5-trimethoxybenzyl)cycloheptanamine hydrobromide

  • MDL number MFCD13186419
  • Cas Number : 1609407-42-2
  • Molecular Weight : 374.3131

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data HBr
Storage temp. Room Temperature
Shipping Temperature Room Temperature
Country of Origin Russia
Product Link https://www.hit2lead.com/building-blocks/5545416

Safety Information

MSDS https://www.hit2lead.com/sds/5545416_sds.pdf
Availability details for
Quantity
  • Availability
  • Add to Cart