CH4424851596 ChemBridge Corporation

N-(3,4,5-trimethoxybenzyl)cyclopentanamine hydrochloride

  • MDL number MFCD06800495
  • Cas Number : 1052525-88-8
  • Molecular Weight : 301.809

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data HCl
Storage temp. Room Temperature
Shipping Temperature Room Temperature
Country of Origin Russia
Product Link https://www.hit2lead.com/building-blocks/5545549

Safety Information

MSDS https://www.hit2lead.com/sds/5545549_sds.pdf
Availability details for
Quantity
  • Availability
  • Add to Cart