CH4424851605 ChemBridge Corporation

N-(4-methoxy-2,5-dimethylbenzyl)-2-phenylethanamine hydrobromide

  • MDL number MFCD13186426
  • Cas Number : 1609407-19-3
  • Molecular Weight : 350.2933

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data HBr
Storage temp. Room Temperature
Shipping Temperature Room Temperature
Country of Origin Russia
Product Link https://www.hit2lead.com/building-blocks/5547245

Safety Information

MSDS https://www.hit2lead.com/sds/5547245_sds.pdf
Availability details for
Quantity
  • Availability
  • Add to Cart