CH4424851607 ChemBridge Corporation

N-(1,3-benzodioxol-5-ylmethyl)-3-pentanamine hydrobromide

  • MDL number MFCD13186428
  • Cas Number : 1609407-98-8
  • Molecular Weight : 302.2074

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data HBr
Storage temp. Room Temperature
Shipping Temperature Room Temperature
Country of Origin Russia
Product Link https://www.hit2lead.com/building-blocks/5547279

Safety Information

MSDS https://www.hit2lead.com/sds/5547279_sds.pdf
Availability details for
Quantity
  • Availability
  • Add to Cart