CH4424851609 ChemBridge Corporation

N-[(4-methoxy-1-naphthyl)methyl]cycloheptanamine hydrobromide

  • MDL number MFCD13186430
  • Cas Number : 1609404-33-2
  • Molecular Weight : 364.3198

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data HBr
Storage temp. Room Temperature
Shipping Temperature Room Temperature
Country of Origin Russia
Product Link https://www.hit2lead.com/building-blocks/5547595

Safety Information

MSDS https://www.hit2lead.com/sds/5547595_sds.pdf
Availability details for
Quantity
  • Availability
  • Add to Cart