CH4424851611 ChemBridge Corporation

N-(5-bromo-2-methoxybenzyl)cyclopentanamine hydrobromide

  • MDL number MFCD13186432
  • Cas Number : 1609404-13-8
  • Molecular Weight : 365.1041

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data HBr
Storage temp. Room Temperature
Shipping Temperature Room Temperature
Country of Origin Russia
Product Link https://www.hit2lead.com/building-blocks/5548045

Safety Information

MSDS https://www.hit2lead.com/sds/5548045_sds.pdf
Availability details for
Quantity
  • Availability
  • Add to Cart