CH4424851618 ChemBridge Corporation

[2-(4-methoxyphenyl)ethyl][4-(methylthio)benzyl]amine hydrochloride

  • MDL number MFCD07110502
  • Cas Number : 1158499-87-6
  • Molecular Weight : 323.8807

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data HCl
Storage temp. RT
Physical Form solid
Shipping Temperature RT
Country of Origin Russia
Product Link https://www.hit2lead.com/building-blocks/5550554

Safety Information

MSDS https://www.hit2lead.com/sds/5550554_sds.pdf
Availability details for
Quantity
  • Availability
  • Add to Cart