CH4424851621 ChemBridge Corporation

(2,5-difluorobenzyl)(4-methoxybenzyl)amine hydrobromide

  • MDL number MFCD13186440
  • Cas Number : 1609403-69-1
  • Molecular Weight : 344.1944

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data HBr
Storage temp. Room Temperature
Shipping Temperature Room Temperature
Country of Origin Russia
Product Link https://www.hit2lead.com/building-blocks/5551609

Safety Information

MSDS https://www.hit2lead.com/sds/5551609_sds.pdf
Availability details for
Quantity
  • Availability
  • Add to Cart